Anti-VWF Mouse Monoclonal Antibody [clone: 1G1]

Proveedor: OriGene
CF803259
ORIGCF803259EA 764 EUR
ORIGCF803259
Anti-VWF Mouse Monoclonal Antibody [clone: 1G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR