Anti-NUMB Mouse Monoclonal Antibody [clone: 4C6]

Proveedor: OriGene
CF802157
ORIGCF802157EA 764 EUR
ORIGCF802157
Anti-NUMB Mouse Monoclonal Antibody [clone: 4C6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NUMB
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR