Anti-VSNL1 Mouse Monoclonal Antibody [clone: 5C8]

Proveedor: OriGene
CF802091
ORIGCF802091EA 764 EUR
ORIGCF802091
Anti-VSNL1 Mouse Monoclonal Antibody [clone: 5C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VSNL1
Clonality: Monoclonal
Clone: 5C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR