Anti-KLK8 Mouse Monoclonal Antibody [clone: 11F7]

Proveedor: OriGene
CF801387
ORIGCF801387EA 764 EUR
ORIGCF801387
Anti-KLK8 Mouse Monoclonal Antibody [clone: 11F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 11F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR