Anti-RBPMS Mouse Monoclonal Antibody [clone: 1C11]

Proveedor: OriGene
CF800217
ORIGCF800217EA 786 EUR
ORIGCF800217
Anti-RBPMS Mouse Monoclonal Antibody [clone: 1C11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: RBPMS
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR