Anti-RBPMS Mouse Monoclonal Antibody [clone: 4F2]

Proveedor: OriGene
CF800207
ORIGCF800207EA 786 EUR
ORIGCF800207
Anti-RBPMS Mouse Monoclonal Antibody [clone: 4F2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RBPMS
Clonality: Monoclonal
Clone: 4F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR