Anti-VWF Mouse Monoclonal Antibody [clone: 2H9]

Proveedor: OriGene
CF803497
ORIGCF803497EA 764 EUR
ORIGCF803497
Anti-VWF Mouse Monoclonal Antibody [clone: 2H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 2H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR