Anti-IL1RN Mouse Monoclonal Antibody [clone: 4E8]

Proveedor: OriGene
CF803484
ORIGCF803484EA 764 EUR
ORIGCF803484
Anti-IL1RN Mouse Monoclonal Antibody [clone: 4E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: IL1RN
Clonality: Monoclonal
Clone: 4E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR