Anti-SYNC Mouse Monoclonal Antibody [clone: 1E7]

Proveedor: OriGene
CF803479
ORIGCF803479EA 764 EUR
ORIGCF803479
Anti-SYNC Mouse Monoclonal Antibody [clone: 1E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SYNC
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR