Anti-NME1 Mouse Monoclonal Antibody [clone: 7F2]

Proveedor: OriGene
CF801264
ORIGCF801264EA 764 EUR
ORIGCF801264
Anti-NME1 Mouse Monoclonal Antibody [clone: 7F2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: 7F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR