Anti-NBN Mouse Monoclonal Antibody [clone: 3A9]

Proveedor: OriGene
CF801251
ORIGCF801251EA 764 EUR
ORIGCF801251
Anti-NBN Mouse Monoclonal Antibody [clone: 3A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: 3A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR