Anti-CDKN2C Mouse Monoclonal Antibody [clone: 5G1]

Proveedor: OriGene
CF801230
ORIGCF801230EA 764 EUR
ORIGCF801230
Anti-CDKN2C Mouse Monoclonal Antibody [clone: 5G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDKN2C
Clonality: Monoclonal
Clone: 5G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR