Anti-SKIL Mouse Monoclonal Antibody [clone: 2C9]

Proveedor: OriGene
CF800172
ORIGCF800172EA 764 EUR
ORIGCF800172
Anti-SKIL Mouse Monoclonal Antibody [clone: 2C9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: SKIL
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR