Anti-FOXI1 Mouse Monoclonal Antibody [clone: 1D4]

Proveedor: OriGene
CF800144
ORIGCF800144EA 764 EUR
ORIGCF800144
Anti-FOXI1 Mouse Monoclonal Antibody [clone: 1D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FOXI1
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR