Anti-ZNF286A Mouse Monoclonal Antibody [clone: 3D6]

Proveedor: OriGene
CF803383
ORIGCF803383EA 786 EUR
ORIGCF803383
Anti-ZNF286A Mouse Monoclonal Antibody [clone: 3D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZNF286A
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR