Anti-BCL6 Mouse Monoclonal Antibody [clone: 11D8]

Proveedor: OriGene
CF803345
ORIGCF803345EA 764 EUR
ORIGCF803345
Anti-BCL6 Mouse Monoclonal Antibody [clone: 11D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BCL6
Clonality: Monoclonal
Clone: 11D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR