Anti-TSC1 Mouse Monoclonal Antibody [clone: 5A8]

Proveedor: OriGene
CF801951
ORIGCF801951EA 764 EUR
ORIGCF801951
Anti-TSC1 Mouse Monoclonal Antibody [clone: 5A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TSC1
Clonality: Monoclonal
Clone: 5A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR