Anti-CD5 Mouse Monoclonal Antibody [clone: 3H4]

Proveedor: OriGene
CF801940
ORIGCF801940EA 764 EUR
ORIGCF801940
Anti-CD5 Mouse Monoclonal Antibody [clone: 3H4]
Anticuerpos
Recommended Dilutions: Flow Cytometry: 1:50

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR