Anti-KRAS Mouse Monoclonal Antibody [clone: 5H6]

Proveedor: OriGene
CF801224
ORIGCF801224EA 764 EUR
ORIGCF801224
Anti-KRAS Mouse Monoclonal Antibody [clone: 5H6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KRAS
Clonality: Monoclonal
Clone: 5H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR