Anti-VIM Mouse Monoclonal Antibody [clone: 7F5]

Proveedor: OriGene
CF801222
ORIGCF801222EA 764 EUR
ORIGCF801222
Anti-VIM Mouse Monoclonal Antibody [clone: 7F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 7F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR