Anti-NBN Mouse Monoclonal Antibody [clone: 1G1]

Proveedor: OriGene
CF801220
ORIGCF801220EA 764 EUR
ORIGCF801220
Anti-NBN Mouse Monoclonal Antibody [clone: 1G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR