Anti-CDKN2C Mouse Monoclonal Antibody [clone: 2E1]

Proveedor: OriGene
CF801184
ORIGCF801184EA 764 EUR
ORIGCF801184
Anti-CDKN2C Mouse Monoclonal Antibody [clone: 2E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDKN2C
Clonality: Monoclonal
Clone: 2E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR