Anti-SENP2 Mouse Monoclonal Antibody [clone: 3E7]

Proveedor: OriGene
CF800127
ORIGCF800127EA 764 EUR
ORIGCF800127
Anti-SENP2 Mouse Monoclonal Antibody [clone: 3E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR