Anti-ZNF365 Mouse Monoclonal Antibody [clone: 4C6]

Proveedor: OriGene
ORIGCF800113EA 764 EUR
ORIGCF800113
Anti-ZNF365 Mouse Monoclonal Antibody [clone: 4C6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZNF365
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR