Anti-PSMA6 Mouse Monoclonal Antibody [clone: 3B8]

Proveedor: OriGene
CF800107
ORIGCF800107EA 764 EUR
ORIGCF800107
Anti-PSMA6 Mouse Monoclonal Antibody [clone: 3B8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: PSMA6
Clonality: Monoclonal
Clone: 3B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR