Anti-PYCARD Mouse Monoclonal Antibody [clone: 1H6]

Proveedor: OriGene
ORIGCF800099EA 764 EUR
ORIGCF800099
Anti-PYCARD Mouse Monoclonal Antibody [clone: 1H6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PYCARD
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR