Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 1G6]

Proveedor: OriGene
CF800089
ORIGCF800089EA 764 EUR
ORIGCF800089
Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 1G6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: UQCRC1
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR