Anti-CDH2 Mouse Monoclonal Antibody [clone: 1C5]

Proveedor: OriGene
CF803191
ORIGCF803191EA 764 EUR
ORIGCF803191
Anti-CDH2 Mouse Monoclonal Antibody [clone: 1C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR