Anti-RIPK3 Mouse Monoclonal Antibody [clone: 2F12]

Proveedor: OriGene
CF803171
ORIGCF803171EA 786 EUR
ORIGCF803171
Anti-RIPK3 Mouse Monoclonal Antibody [clone: 2F12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RIPK3
Clonality: Monoclonal
Clone: 2F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR