Anti-VSNL1 Mouse Monoclonal Antibody [clone: 1F10]

Proveedor: OriGene
ORIGCF801902EA 764 EUR
ORIGCF801902
Anti-VSNL1 Mouse Monoclonal Antibody [clone: 1F10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VSNL1
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR