Anti-WFDC2 Mouse Monoclonal Antibody [clone: 1E12]

Proveedor: OriGene
CF801136
ORIGCF801136EA 764 EUR
ORIGCF801136
Anti-WFDC2 Mouse Monoclonal Antibody [clone: 1E12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: WFDC2
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR