Anti-VIM Mouse Monoclonal Antibody [clone: 3G4]

Proveedor: OriGene
CF801122
ORIGCF801122EA 764 EUR
ORIGCF801122
Anti-VIM Mouse Monoclonal Antibody [clone: 3G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR