Anti-C1R Mouse Monoclonal Antibody [clone: 5A11]

Proveedor: OriGene
ORIGCF800084EA 764 EUR
ORIGCF800084
Anti-C1R Mouse Monoclonal Antibody [clone: 5A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: C1R
Clonality: Monoclonal
Clone: 5A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR