Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 1A3]

Proveedor: OriGene
CF800070
ORIGCF800070EA 764 EUR
ORIGCF800070
Anti-UQCRC1 Mouse Monoclonal Antibody [clone: 1A3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: UQCRC1
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR