Anti-SAP30BP Mouse Monoclonal Antibody [clone: 3A11]

Proveedor: OriGene
CF803105
ORIGCF803105EA 764 EUR
ORIGCF803105
Anti-SAP30BP Mouse Monoclonal Antibody [clone: 3A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SAP30BP
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR