Anti-SAP30BP Mouse Monoclonal Antibody [clone: 4A9]

Proveedor: OriGene
CF803084
ORIGCF803084EA 764 EUR
ORIGCF803084
Anti-SAP30BP Mouse Monoclonal Antibody [clone: 4A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SAP30BP
Clonality: Monoclonal
Clone: 4A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR