Anti-TNFRSF9 Mouse Monoclonal Antibody [clone: 5H5]

Proveedor: OriGene
CF801748
ORIGCF801748EA 764 EUR
ORIGCF801748
Anti-TNFRSF9 Mouse Monoclonal Antibody [clone: 5H5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TNFRSF9
Clonality: Monoclonal
Clone: 5H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR