Anti-DOCK8 Mouse Monoclonal Antibody [clone: 1E11]

Proveedor: OriGene
CF801739
ORIGCF801739EA 764 EUR
ORIGCF801739
Anti-DOCK8 Mouse Monoclonal Antibody [clone: 1E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DOCK8
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR