Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 3B5]

Proveedor: OriGene
CF801726
ORIGCF801726EA 764 EUR
ORIGCF801726
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 3B5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR