Anti-ABCB1 Mouse Monoclonal Antibody [clone: 1F8]

Proveedor: OriGene
CF801012
ORIGCF801012EA 764 EUR
ORIGCF801012
Anti-ABCB1 Mouse Monoclonal Antibody [clone: 1F8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: ABCB1
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR