Anti-CD99 Mouse Monoclonal Antibody [clone: 1F1]

Proveedor: OriGene
CF800963
ORIGCF800963EA 764 EUR
ORIGCF800963
Anti-CD99 Mouse Monoclonal Antibody [clone: 1F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR