Anti-DPPA4 Mouse Monoclonal Antibody [clone: 1C1]

Proveedor: OriGene
CF800025
ORIGCF800025EA 786 EUR
ORIGCF800025
Anti-DPPA4 Mouse Monoclonal Antibody [clone: 1C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: DPPA4
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR