Anti-C17orf37 Mouse Monoclonal Antibody [clone: 4D4]

Proveedor: OriGene
CF800015
ORIGCF800015EA 764 EUR
ORIGCF800015
Anti-C17orf37 Mouse Monoclonal Antibody [clone: 4D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: C17orf37
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR