Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 2C2]

Proveedor: OriGene
CF801639
ORIGCF801639EA 764 EUR
ORIGCF801639
Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 2C2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PIK3C2A
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR