Anti-PCNA Mouse Monoclonal Antibody [clone: 3F11]

Proveedor: OriGene
CF800915
ORIGCF800915EA 786 EUR
ORIGCF800915
Anti-PCNA Mouse Monoclonal Antibody [clone: 3F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR