Anti-CTNND1 Mouse Monoclonal Antibody [clone: 2F8]

Proveedor: OriGene
CF800905
ORIGCF800905EA 764 EUR
ORIGCF800905
Anti-CTNND1 Mouse Monoclonal Antibody [clone: 2F8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CTNND1
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR