Anti-PCNA Mouse Monoclonal Antibody [clone: 5B10]

Proveedor: OriGene
CF800879
ORIGCF800879EA 786 EUR
ORIGCF800879
Anti-PCNA Mouse Monoclonal Antibody [clone: 5B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 5B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR