Anti-PCNA Mouse Monoclonal Antibody [clone: 4G3]

Proveedor: OriGene
CF800876
ORIGCF800876EA 786 EUR
ORIGCF800876
Anti-PCNA Mouse Monoclonal Antibody [clone: 4G3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR