Anti-SPG7 Mouse Monoclonal Antibody [clone: 1D10]

Proveedor: OriGene
CF800002
ORIGCF800002EA 764 EUR
ORIGCF800002
Anti-SPG7 Mouse Monoclonal Antibody [clone: 1D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SPG7
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR