Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 1F11]

Proveedor: OriGene
CF507305
ORIGCF507305EA 786 EUR
ORIGCF507305
Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 1F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150

Type: Primary
Antigen: ANKMY2
Clonality: Monoclonal
Clone: 1F11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR